accessibility menu, dialog, popup

UserName

CAS RN 161337-67-3

FFFSOOOFFFSOOOFFFSOOOFFFSOOOH2OHf

CAS RN 161337-67-3

IUPAC Name: hafnium(4+) hydrate tetratrifluoromethanesulfonate
Synonyms: hafnium(4+) hydrate tetratriflate
Molecular Weight (g/mol): 792.76
Molecular Formula: C4H2F12HfO13S4
InChi Key: XQJOFTWVKHJLHA-UHFFFAOYSA-J
SMILES: O.[Hf+4].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)