missing translation for 'onlineSavingsMsg'
Learn More

Hafnium trifluoromethanesulfonate, 98%

Catalog No. AAL1968814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1968814 25 g
AAL1968806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1968814 Supplier Thermo Scientific Chemicals Supplier No. L1968814
Only null left

CAS: 161337-67-3 | C4H2F12HfO13S4 | 792.758 g/mol

Hafnium trifluoromethanesulfonate is a recyclable catalyst for the mononitration of o-nitrotoluene. It serves as a catalyst for homogeneous methoxycarbonylation and hydrocarboxylation reactions of phenylacetylene. It is also used as a reactant for direct Friedel-Crafts reactions of chromene hemiacetals, aminomethylation reactions under Lewis acidic conditions and prins-type cyclization reactions. It is also involved in the direct polycondensation of lactic acid. Further, it is used in cationic benzylation reactions, chemoselective thioacetalization and transthioacetalization of carbonyl compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hafnium trifluoromethanesulfonate is a recyclable catalyst for the mononitration of o-nitrotoluene. It serves as a catalyst for homogeneous methoxycarbonylation and hydrocarboxylation reactions of phenylacetylene. It is also used as a reactant for direct Friedel-Crafts reactions of chromene hemiacetals, aminomethylation reactions under Lewis acidic conditions and prins-type cyclization reactions. It is also involved in the direct polycondensation of lactic acid. Further, it is used in cationic benzylation reactions, chemoselective thioacetalization and transthioacetalization of carbonyl compounds.

Solubility
Soluble in methyl cyanide, dicholomethane, dichloroethane, nitromethane, dioxane, terahydrofuran and toluene.

Notes
Incompatible with strong oxidizing agents. Moisture sensitive.

Chemical Identifiers

CAS 161337-67-3
Molecular Formula C4H2F12HfO13S4
Molecular Weight (g/mol) 792.758
MDL Number MFCD01321255
InChI Key XQJOFTWVKHJLHA-UHFFFAOYSA-J
Synonym hafnium iv trifluoromethanesulfonate hydrate, hafnium tetrakis trifluoromethanesulfonate , hydrate
PubChem CID 71433309
IUPAC Name hafnium(4+);trifluoromethanesulfonate;hydrate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].O.[Hf+4]

Specifications

Melting Point >350°C
Linear Formula (CF3SO3)4Hf
Quantity 25 g
Sensitivity Hygroscopic
Solubility Information Soluble in methyl cyanide,dicholomethane,dichloroethane,nitromethane,dioxane,terahydrofuran and toluene.
Formula Weight 774.75
Percent Purity 98%
Chemical Name or Material Hafnium trifluoromethanesulfonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only