accessibility menu, dialog, popup

UserName

CH3SiCH3CH3I

CAS RN 16029-98-4

IUPAC Name: iodotrimethylsilane
Synonyms: trimethylsilyl iodide
Molecular Weight (g/mol): 200.09
Molecular Formula: C3H9ISi
InChi Key: CSRZQMIRAZTJOY-UHFFFAOYSA-N
SMILES: C[Si](C)(C)I
Molecular Weight (g/mol) 
  • (6)
  • (5)
Percent Purity 
  • (2)
  • (4)
  • (1)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (2)
  • (9)
  • (6)
  • (5)
  • (2)
  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
  • (7)