accessibility menu, dialog, popup

UserName

CAS RN 1594092-37-1

H3CNNHNNNNNH3COCH3

CAS RN 1594092-37-1

IUPAC Name: N-[2-methoxy-4-(4-methylpiperazin-1-yl)phenyl]-4-{1-methyl-1H-pyrrolo[2,3-c]pyridin-3-yl}pyrimidin-2-amine
Synonyms: N-[2-methoxy-4-(4-methylpiperazin-1-yl)phenyl]-4-{1-methylpyrrolo[2,3-c]pyridin-3-yl}pyrimidin-2-amine
Molecular Weight (g/mol): 429.53
Molecular Formula: C24H27N7O
InChi Key: ZYVXTMKTGDARKR-UHFFFAOYSA-N
SMILES: COC1=C(NC2=NC=CC(=N2)C2=CN(C)C3=CN=CC=C23)C=CC(=C1)N1CCN(C)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results