accessibility menu, dialog, popup

UserName

CAS RN 15923-42-9

IUPAC Name: 6-methyl-1H,2H,3H,4H,5H,6H-azepino[4,5-b]indol-6-ium chloride
Synonyms: 6-methyl-1H,2H,3H,4H,5H,6H-azepino[4,5-b]indol-6-ium chloride
Molecular Weight (g/mol): 236.74
Molecular Formula: C13H17ClN2
InChi Key: SAIGGEUWSYESTR-UHFFFAOYNA-N
SMILES: [Cl-].C[NH+]1C2=C(CCNCC2)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results