missing translation for 'onlineSavingsMsg'
Learn More

PNU 22394 hydrochloride, Tocris Bioscience™

Catalog No. 220110
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
220110 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 220110 Supplier SPEX CertiPrep Group LLC Supplier No. 220110

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

5-HT2C agonist and 5-HT2A/2B partial agonist.

Potent 5-HT2C agonist and partial 5-HT2A/5-HT2B agonist. Non-selective between 5-HT2 receptor subtypes (Ki values are 18, 18 and 66 nM for human recombinant 5-HT2C, 5-HT2A and 5-HT2B receptors respectively). Reduces food intake in rats following subcutaneous administration and displays anorexigenic effects in humans.

Chemical Identifiers

CAS 15923-42-9
Molecular Formula C13H17ClN2
Molecular Weight (g/mol) 236.743
InChI Key SAIGGEUWSYESTR-UHFFFAOYSA-N
Synonym pnu 22394 hydrochloride, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indole hydrochloride, 6-methyl-1h,2h,3h,4h,5h-azepino 4,5-b indole hydrochloride, 6-methyl-1,2,3,4,5,6-hexahydroazepino 4,5-b indol-6-ium chloride, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indol, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indole-hydrochloride
PubChem CID 16759162
IUPAC Name 6-methyl-2,3,4,5-tetrahydro-1H-azepino[4,5-b]indole;hydrochloride
SMILES CN1C2=C(CCNCC2)C3=CC=CC=C31.Cl

Specifications

Quantity 10 mg
Formula Weight 236.74
Percent Purity >99%
Chemical Name or Material PNU 22394 hydrochloride
Recommended Storage Desiccate at +4°C