accessibility menu, dialog, popup

UserName

CAS RN 15905-32-5

IUPAC Name: 3',6'-dihydroxy-2',4',5',7'-tetraiodo-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: erythrosine|E127 erythrosine|erythrosine, phenolic
Molecular Weight (g/mol): 835.9
Molecular Formula: C20H8I4O5
InChi Key: OALHHIHQOFIMEF-UHFFFAOYSA-N
SMILES: OC1=C(I)C2=C(C=C1I)C1(OC(=O)C3=C1C=CC=C3)C1=CC(I)=C(O)C(I)=C1O2
Quantity 
  • (1)
  • (2)
  • (2)
Color 
  • (3)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)