accessibility menu, dialog, popup

UserName

BFFFFHOOH

CAS RN 157834-21-4

IUPAC Name: [2-fluoro-3-(trifluoromethyl)phenyl]boronic acid
Synonyms: 2-fluoro-3-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 207.92
Molecular Formula: C7H5BF4O2
InChi Key: PKARAEQMGHJHMY-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C(=CC=C1)C(F)(F)F
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results