missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F10061G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10061G Supplier TCI America Supplier No. F10061G

Specifications

Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 157834-21-4
Melting Point 58°C
Color White
Molecular Formula C7H5BF4O2
Quantity 1 g
MDL Number MFCD07784336
Synonym 2-fluoro-3-trifluoromethyl phenylboronic acid, 2-fluoro-3-trifluoromethyl benzeneboronic acid, 2-fluoro-3-trifluoromethyl phenyl boronic acid, 2-fluoro-3-trifluoromethylphenylboronic acid, 2-fluoro-3-trifluoromethyl phenylboron, 2-fluoro-3-trifluoromethyl phenylboronicacid, boronic acid, 2-fluoro-3-trifluoromethyl phenyl, boronic acid,b-2-fluoro-3-trifluoromethyl phenyl, boronic acid, b-2-fluoro-3-trifluoromethyl phenyl
InChI Key PKARAEQMGHJHMY-UHFFFAOYSA-N
SMILES B(C1=C(C(=CC=C1)C(F)(F)F)F)(O)O
IUPAC Name [2-fluoro-3-(trifluoromethyl)phenyl]boronic acid
Molecular Weight (g/mol) 207.919
PubChem CID 16217169
Formula Weight 207.92
Physical Form Crystalline Powder
Show More Show Less