accessibility menu, dialog, popup

UserName

CH3(S)H2COO

CAS RN 1575-74-2

IUPAC Name: (2S)-2-methylpent-4-enoate
Synonyms: (2S)-2-methylpent-4-enoate
Molecular Weight (g/mol): 113.14
Molecular Formula: C6H9O2
InChi Key: HVRZYSHVZOELOH-YFKPBYRVSA-M
SMILES: C[C@@H](CC=C)C([O-])=O
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)