missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-pentenoic acid, 98%

Catalog No. AAA1129718
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1129718 50 g
AAA1129709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1129718 Supplier Thermo Scientific Chemicals Supplier No. A1129718
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1575-74-2 | C6H10O2 | 114.144 g/mol

2-Methyl-4-pentenoic acid is used as raw material for fruit flavor or it is an enhancer of fruity fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-4-pentenoic acid is used as raw material for fruit flavor or it is an enhancer of fruity fragrances.

Solubility
Slightly soluble in water

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1575-74-2
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00015570
InChI Key HVRZYSHVZOELOH-UHFFFAOYSA-N
Synonym 2-methyl-4-pentenoic acid, 4-pentenoic acid, 2-methyl, 2-methylpent-4-en-1-oic acid, fema no. 3511, 2-methyl-4-pentenoicacid, methylallylacetic acid, acmc-209dg9, 2-methyl-pent-4-enoic acid, hvrzyshvzoeloh-uhfffaoysa, 2-methyl-4-pentenoic acid, fg
PubChem CID 549519
IUPAC Name 2-methylpent-4-enoic acid
SMILES CC(CC=C)C(=O)O

Specifications

Density 0.95
Boiling Point 195°C
Flash Point 107°C (224°F)
Odor Characteristic
Refractive Index 1.431
Quantity 50 g
UN Number UN3265
Beilstein 1720987
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water
Formula Weight 114.15
Percent Purity 98%
Chemical Name or Material 2-Methyl-4-pentenoic acid
Show More Show Less
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 216-404-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only