accessibility menu, dialog, popup

UserName

CAS RN 1574-41-0

CH3CH2(Z)

CAS RN 1574-41-0

IUPAC Name: (3Z)-penta-1,3-diene
Synonyms: 1,3-pentadiene (Z)
Molecular Weight (g/mol): 68.12
Molecular Formula: C5H8
InChi Key: PMJHHCWVYXUKFD-PLNGDYQASA-N
SMILES: C\C=C/C=C
Percent Purity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results