accessibility menu, dialog, popup

UserName

CAS RN 157212-55-0

H2OCH3OONHSOOCH3H3CCH3NNOOHNN

CAS RN 157212-55-0

IUPAC Name: 4-tert-butyl-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-[2,2'-bipyrimidin]-4-yl]benzene-1-sulfonamide hydrate
Synonyms: bosentan hydrate
Molecular Weight (g/mol): 569.63
Molecular Formula: C27H31N5O7S
InChi Key: SXTRWVVIEPWAKM-UHFFFAOYSA-N
SMILES: O.COC1=CC=CC=C1OC1=C(NS(=O)(=O)C2=CC=C(C=C2)C(C)(C)C)N=C(N=C1OCCO)C1=NC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results