accessibility menu, dialog, popup

UserName

CAS RN 1568-70-3

IUPAC Name: 4-methoxy-2-nitrophenol
Synonyms: 4-methoxy-2-nitrophenol
Molecular Weight (g/mol): 169.14
Molecular Formula: C7H7NO4
InChi Key: YBUGOACXDPDUIR-UHFFFAOYSA-N
SMILES: COC1=CC=C(O)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results