accessibility menu, dialog, popup

UserName

CAS RN 15629-92-2

IUPAC Name: [3-(diphenylphosphanyl)propyl]diphenylphosphane; dichloronickelbis(ylium)
Synonyms: [3-(diphenylphosphanyl)propyl]diphenylphosphane; dichloronickelbis(ylium)
Molecular Weight (g/mol): 542.04
Molecular Formula: C27H26Cl2NiP2
InChi Key: REQVOKPBMSCTNI-UHFFFAOYSA-L
SMILES: Cl[Ni++]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)