missing translation for 'onlineSavingsMsg'
Learn More

[1,3-Bis(diphenylphosphino)propane]nickel(II) Dichloride 98.0+%, TCI America™

Catalog No. B13135G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13135G 5 g
B131325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13135G Supplier TCI America Supplier No. B13135G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15629-92-2
Molecular Formula C27H26Cl2NiP2
Molecular Weight (g/mol) 542.04
MDL Number MFCD00015318
InChI Key REQVOKPBMSCTNI-UHFFFAOYSA-L
Synonym [1,3-Bis(diphenylphosphino)propane]dichloronickel(II), Dichloro[1,3-bis(diphenylphosphino)propane]nickel(II)
PubChem CID 131675641
IUPAC Name [3-(diphenylphosphanyl)propyl]diphenylphosphane; dichloronickelbis(ylium)
SMILES Cl[Ni++]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 213°C
Quantity 5 g
Formula Weight 542.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [1,3-Bis(diphenylphosphino)propane]nickel(II) Dichloride