missing translation for 'onlineSavingsMsg'
Learn More

[1,3-Bis(diphenylphosphino)propane]nickel(II) chloride, 99%, Thermo Scientific™

Catalog No. AC291590250
Change view
Click to view available options
Glass bottle; 25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC291590250 Glass bottle; 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC291590250 Supplier Thermo Scientific Chemicals Supplier No. 291590250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 15629-92-2
CAS Min % 98.5
CAS Max % 100.0
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C27H26Cl2NiP2
Quantity 25g
MDL Number MFCD00015318
Fieser 06,50; 15,122
Solubility Information Solubility in water: insoluble
InChI Key BILYTLYIOVUVSU-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)P(CCCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl.[Ni]
IUPAC Name 3-diphenylphosphanylpropyl(diphenyl)phosphane;nickel;dihydrochloride
Molecular Weight (g/mol) 542.04
PubChem CID 131675641
Formula Weight 542.04
Percent Purity 99%
Chemical Name or Material [1,3-Bis(diphenylphosphino)propane]nickel(II) chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON

RUO – Research Use Only