accessibility menu, dialog, popup

UserName

CH3SiClClCl

CAS RN 1558-33-4

IUPAC Name: dichloro(chloromethyl)methylsilane
Synonyms: dichloro(chloromethyl)methylsilane
Molecular Weight (g/mol): 163.5
Molecular Formula: C2H5Cl3Si
InChi Key: JAYBZWYBCUJLNQ-UHFFFAOYSA-N
SMILES: C[Si](Cl)(Cl)CCl
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)