missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl(dichloro)methylsilane 97.0+%, TCI America™

Catalog No. C135825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C135825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C135825G Supplier TCI America Supplier No. C135825G

Chemical Identifiers

CAS 1558-33-4
Molecular Formula C2H5Cl3Si
Molecular Weight (g/mol) 163.497
MDL Number MFCD00000874
InChI Key JAYBZWYBCUJLNQ-UHFFFAOYSA-N
Synonym dichloro chloromethyl methylsilane, silane, dichloro chloromethyl methyl, ch3sicl2 ch2cl, chloromethyl methyldichlorosilane, chloromethylmethyldichlorosilane, unii-9p7n047n2s, chloromethyl dichloro methylsilane, dichloro chloromethyl methyl silane, silane cmm1, chloromethyldichloro methyl silane
PubChem CID 73788
IUPAC Name dichloro-(chloromethyl)-methylsilane
SMILES C[Si](CCl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 122°C
Quantity 25 g
UN Number 3362
Formula Weight 163.50
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloromethyl(dichloro)methylsilane
EINECSNumber (2)-2047
RTECSNumber VV2160000
TSCA Yes
Recommended Storage Refrigerator