missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(chloromethyl)methylsilane, 95%

Catalog No. AC313371000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC313371000 100 g Glass bottle
AC313370250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC313371000 Supplier Thermo Scientific Chemicals Supplier No. 313371000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1558-33-4 | C2H5Cl3Si | 163.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1558-33-4
Molecular Formula C2H5Cl3Si
Molecular Weight (g/mol) 163.51
InChI Key JAYBZWYBCUJLNQ-UHFFFAOYSA-N
Synonym dichloro chloromethyl methylsilane, silane, dichloro chloromethyl methyl, ch3sicl2 ch2cl, chloromethyl methyldichlorosilane, chloromethylmethyldichlorosilane, unii-9p7n047n2s, chloromethyl dichloro methylsilane, dichloro chloromethyl methyl silane, silane cmm1, chloromethyldichloro methyl silane
PubChem CID 73788
IUPAC Name dichloro-(chloromethyl)-methylsilane
SMILES C[Si](CCl)(Cl)Cl

Specifications

Color Colorless to Straw
Density 1.2600g/mL
Boiling Point 121.0°C to 122.0°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4480 to 1.4510
Linear Formula ClCH2Si(Cl2)CH3
Quantity 100 g
Specific Gravity 1.26
Solubility Information Solubility in water: reacts
Formula Weight 163.51
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Dichloro(chloromethyl)methylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
Flammable liquid and vapor.
Harmful to aquatic life with long lasting effects.
Reacts violently with water.
Corrosive to the respiratory tract
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
EINECSNumber 216-319-5

RUO – Research Use Only