CAS RN 15567-09-6
CAS RN 15567-09-6
IUPAC Name:
butan-1-amine hydrobromide
Synonyms:
n-butylamine hydrobromide
Molecular Weight (g/mol):
154.05
Molecular Formula:
C4H12BrN
InChi Key:
SXGBREZGMJVYRL-UHFFFAOYSA-N
SMILES:
Br.CCCCN
1
–
1
of
1
results
1
Butylamine Hydrobromide 98.0+%, TCI America™
CAS: 15567-09-6 Molecular Formula: C4H12BrN Molecular Weight (g/mol): 154.051 InChI Key: SXGBREZGMJVYRL-UHFFFAOYSA-N Synonym: Butylammonium Bromide PubChem CID: 12199058 IUPAC Name: butan-1-amine;hydrobromide SMILES: CCCCN.Br