missing translation for 'onlineSavingsMsg'
Learn More

Butylamine Hydrobromide 98.0+%, TCI America™

$73.16 - $221.39

Chemical Identifiers

CAS 15567-09-6
Molecular Formula C4H12BrN
Molecular Weight (g/mol) 154.051
InChI Key SXGBREZGMJVYRL-UHFFFAOYSA-N
Synonym Butylammonium Bromide
PubChem CID 12199058
IUPAC Name butan-1-amine;hydrobromide
SMILES CCCCN.Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B51861G
View Documents
TCI America
B51861G
1 g
Each for $73.16
Only null left
 
B51865G
View Documents
TCI America
B51865G
5 g
Each for $221.39
Only null left
 
Description

Description

Specifications

Chemical Identifiers

15567-09-6
154.051
Butylammonium Bromide
butan-1-amine;hydrobromide
C4H12BrN
SXGBREZGMJVYRL-UHFFFAOYSA-N
12199058
CCCCN.Br

Specifications

15567-09-6
C4H12BrN
Butylammonium Bromide
CCCCN.Br
154.051
154.05
Crystalline Powder
White
1 g
SXGBREZGMJVYRL-UHFFFAOYSA-N
butan-1-amine;hydrobromide
12199058
≥98.0% (N,T)
Butylamine Hydrobromide
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents