missing translation for 'onlineSavingsMsg'
Learn More

Dilithium Tetrachlorocopper(II) (ca. 2.5% in Tetrahydrofuran, ca. 0.1mol/L), TCI America™

Catalog No. D4395100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4395100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4395100ML Supplier TCI America Supplier No. D4395100ML
Only null left

Chemical Identifiers

CAS 15489-27-7
Molecular Formula Cl4CuLi2
Molecular Weight (g/mol) 219.226
MDL Number MFCD00011081
InChI Key HCJWWBBBSCXJMS-UHFFFAOYSA-J
Synonym Dilithium Tetrachlorocuprate(II)
PubChem CID 11074879
IUPAC Name dilithium;tetrachlorocopper(2-)
SMILES [Li+].[Li+].Cl[Cu-2](Cl)(Cl)Cl

Specifications

Quantity 100 mL
UN Number 2056
Formula Weight 219.23
Physical Form Liquid
Chemical Name or Material Dilithium Tetrachlorocopper(II) (ca. 2.5% in Tetrahydrofuran, ca. 0.1mol/L)