accessibility menu, dialog, popup

UserName

H3CNOSOO

CAS RN 15448-99-4

IUPAC Name: 2-methyl-2,3-dihydro-1λ⁶,2-benzothiazole-1,1,3-trione
Synonyms: 2-methyl-1λ⁶,2-benzothiazole-1,1,3-trione
Molecular Weight (g/mol): 197.21
Molecular Formula: C8H7NO3S
InChi Key: DDIIAJRLFATEEE-UHFFFAOYSA-N
SMILES: CN1C(=O)C2=CC=CC=C2S1(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results