missing translation for 'onlineSavingsMsg'
Learn More

N-Methylsaccharin 98.0+%, TCI America™

Catalog No. M042725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M042725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M042725G Supplier TCI America Supplier No. M042725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15448-99-4
Molecular Formula C8H7NO3S
Molecular Weight (g/mol) 197.21
MDL Number MFCD00059263
InChI Key DDIIAJRLFATEEE-UHFFFAOYSA-N
PubChem CID 27290
IUPAC Name 2-methyl-2,3-dihydro-1λ⁶,2-benzothiazole-1,1,3-trione
SMILES CN1C(=O)C2=CC=CC=C2S1(=O)=O

Specifications

Melting Point 131°C
Color White
Quantity 25 g
Formula Weight 197.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Methylsaccharin