accessibility menu, dialog, popup

UserName

CAS RN 154071-48-4

OOONOHOOHOOHHONOHOOHO

CAS RN 154071-48-4

IUPAC Name: 2-{[(7'-{[bis(carboxymethyl)amino]methyl}-3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-2'-yl)methyl](carboxymethyl)amino}acetic acid
Synonyms: calcein oftasceina oftasceine oftasceinum 奥荧光素
Molecular Weight (g/mol): 622.54
Molecular Formula: C30H26N2O13
InChi Key: DEGAKNSWVGKMLS-UHFFFAOYSA-N
SMILES: OC(=O)CN(CC(O)=O)CC1=C(O)C=C2OC3=CC(O)=C(CN(CC(O)=O)CC(O)=O)C=C3C3(OC(=O)C4=CC=CC=C34)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results