missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcein (mixture of isomers) [for Fluorometric Determination of Ca], TCI America™
Specifications
Specifications
| Formula Weight | 622.54 |
| Quantity | 5 g |
| Physical Form | Crystal/Powder |
| Chemical Name or Material | Calcein (mixture of isomers) [for Fluorometric Determination of Ca] |
Safety and Handling
| TSCA | Yes |