accessibility menu, dialog, popup

UserName

HClH3CNCH3NN

CAS RN 154-69-8

IUPAC Name: N-benzyl-N-[2-(dimethylamino)ethyl]pyridin-2-amine hydrochloride
Synonyms: tripelennamine hydrochloride
Molecular Weight (g/mol): 291.82
Molecular Formula: C16H22ClN3
InChi Key: FSSICIQKZGUEAE-UHFFFAOYSA-N
SMILES: Cl.CN(C)CCN(CC1=CC=CC=C1)C1=CC=CC=N1
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results