CAS RN 153587-01-0
IUPAC Name:
1-[2-hydroxy-5-(trifluoromethyl)phenyl]-5-(trifluoromethyl)-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms:
1-[2-hydroxy-5-(trifluoromethyl)phenyl]-5-(trifluoromethyl)-3H-1,3-benzodiazol-2-one
Molecular Weight (g/mol):
362.23
Molecular Formula:
C15H8F6N2O2
InChi Key:
YLFMCMWKHSDUCT-UHFFFAOYSA-N
SMILES:
OC1=CC=C(C=C1N1C(=O)NC2=CC(=CC=C12)C(F)(F)F)C(F)(F)F