accessibility menu, dialog, popup

UserName

CAS RN 153531-96-5

OHOOHOOOHHOOHOOHOOOHHOH2OH2OH2OH2OH2OH2OH2OH2OH2OMgMgMg

CAS RN 153531-96-5

IUPAC Name: bis(2-hydroxypropane-1,2,3-tricarboxylic acid) nonahydrate trimagnesium
Synonyms: bis(citric acid) nonahydrate trimagnesium
Molecular Weight (g/mol): 619.3
Molecular Formula: C12H34Mg3O23
InChi Key: YMNRVQRRBHEJPL-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.O.O.O.[Mg].[Mg].[Mg].OC(=O)CC(O)(CC(O)=O)C(O)=O.OC(=O)CC(O)(CC(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results