missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimagnesium Dicitrate Nonahydrate 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 500 g |
| Formula Weight | 451.11 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Trimagnesium Dicitrate Nonahydrate |
Safety and Handling
| EINECSNumber | (2)-1323 |
| RTECSNumber | TZ8552900 |
| TSCA | No |