missing translation for 'onlineSavingsMsg'
Learn More

4'-Propyl-4-biphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. P23871G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P23871G Supplier TCI America Supplier No. P23871G

Specifications

Chemical Name or Material 4′-Propyl-4-biphenylboronic Acid (contains varying amounts of Anhydride)
CAS 153035-56-4
Color White-Yellow
Molecular Formula C15H17BO2
Quantity 1 g
MDL Number MFCD01114670
InChI Key NOQFUISBLHCDSR-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)C2=CC=C(C=C2)CCC)(O)O
IUPAC Name [4-(4-propylphenyl)phenyl]boronic acid
Molecular Weight (g/mol) 240.109
PubChem CID 19042596
Formula Weight 240.11
Physical Form Crystalline Powder
Show More Show Less