CAS RN 1528-53-6
CAS RN 1528-53-6
IUPAC Name:
1,2,4-tripropyl benzene-1,2,4-tricarboxylate
Synonyms:
1,2,4-tripropyl benzene-1,2,4-tricarboxylate
Molecular Weight (g/mol):
336.38
Molecular Formula:
C18H24O6
InChi Key:
QEUYMNVHNSOBRS-UHFFFAOYSA-N
SMILES:
CCCOC(=O)C1=CC(C(=O)OCCC)=C(C=C1)C(=O)OCCC
1
–
1
of
1
results
1
Tripropyl Trimellitate 97.0+%, TCI America™
CAS: 1528-53-6 Molecular Formula: C18H24O6 Molecular Weight (g/mol): 336.384 MDL Number: MFCD00191686 InChI Key: QEUYMNVHNSOBRS-UHFFFAOYSA-N Synonym: 1,2,4-Benzenetricarboxylic Acid Tripropyl Ester, Trimellitic Acid Tripropyl Ester PubChem CID: 15920120 IUPAC Name: tripropyl benzene-1,2,4-tricarboxylate SMILES: CCCOC(=O)C1=CC(=C(C=C1)C(=O)OCCC)C(=O)OCCC