missing translation for 'onlineSavingsMsg'
Learn More

Tripropyl Trimellitate 97.0+%, TCI America™

Catalog No. T144625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T144625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T144625G Supplier TCI America Supplier No. T144625G
Only null left

Chemical Identifiers

CAS 1528-53-6
Molecular Formula C18H24O6
Molecular Weight (g/mol) 336.384
MDL Number MFCD00191686
InChI Key QEUYMNVHNSOBRS-UHFFFAOYSA-N
Synonym 1,2,4-Benzenetricarboxylic Acid Tripropyl Ester, Trimellitic Acid Tripropyl Ester
PubChem CID 15920120
IUPAC Name tripropyl benzene-1,2,4-tricarboxylate
SMILES CCCOC(=O)C1=CC(=C(C=C1)C(=O)OCCC)C(=O)OCCC

Specifications

Color Yellow
Boiling Point 168°C
Quantity 25 g
Formula Weight 336.38
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tripropyl Trimellitate