accessibility menu, dialog, popup

UserName

CH3ONH2OOCH3

CAS RN 1521-39-7

IUPAC Name: 2,3-dimethoxybenzamide
Synonyms: 2,3-dimethoxybenzamide
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: SDYIZAANGZBOSO-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C(N)=O)=C1OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results