missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxybenzamide 98.0+%, TCI America™

Catalog No. D22835G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D22835G Supplier TCI America Supplier No. D22835G
Only null left

Chemical Identifiers

CAS 1521-39-7
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00017571
InChI Key SDYIZAANGZBOSO-UHFFFAOYSA-N
PubChem CID 220089
IUPAC Name 2,3-dimethoxybenzamide
SMILES COC1=CC=CC(C(N)=O)=C1OC

Specifications

Melting Point 94°C
Quantity 5 g
Formula Weight 181.19
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethoxybenzamide