CAS RN 152-47-6
IUPAC Name:
4-amino-N-(3-methoxypyrazin-2-yl)benzene-1-sulfonamide
Synonyms:
longum sulfalene sulfamethoxypyrazine SMP kelfizin kelfizina kelfizine sulfametopyrazine solfametopirazina farmitalia 204/122
Molecular Weight (g/mol):
280.3
Molecular Formula:
C11H12N4O3S
InChi Key:
KXRZBTAEDBELFD-UHFFFAOYSA-N
SMILES:
COC1=NC=CN=C1NS(=O)(=O)C1=CC=C(N)C=C1
1
–
1
of
1
results
1
Sulfalene 98.0+%, TCI America™
CAS: 152-47-6 Molecular Formula: C11H12N4O3S Molecular Weight (g/mol): 280.302 MDL Number: MFCD00437754 InChI Key: KXRZBTAEDBELFD-UHFFFAOYSA-N Synonym: 4-Amino-N-(3-methoxypyrazin-2-yl)benzenesulfonamide, N-(3-Methoxypyrazin-2-yl)sulfanilamide PubChem CID: 9047 ChEBI: CHEBI:32162 IUPAC Name: 4-amino-N-(3-methoxypyrazin-2-yl)benzenesulfonamide SMILES: COC1=NC=CN=C1NS(=O)(=O)C2=CC=C(C=C2)N