accessibility menu, dialog, popup

UserName

H3CNOHNHCl

CAS RN 151257-01-1

IUPAC Name: 2-butyl-1,3-diazaspiro[4.4]non-1-en-4-one hydrochloride
Synonyms: 2-butyl-1,3-diazaspiro[4.4]non-1-en-4-one hydrochloride
Molecular Weight (g/mol): 230.74
Molecular Formula: C11H19ClN2O
InChi Key: WWRHZLCKSVQRBG-UHFFFAOYSA-N
SMILES: Cl.CCCCC1=NC2(CCCC2)C(=O)N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results