missing translation for 'onlineSavingsMsg'
Learn More

2-Butyl-1,3-diazaspiro[4.4]non-1-en-4-one Hydrochloride 98.0+%, TCI America™

Catalog No. B32915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32915G 5 g
B329125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32915G Supplier TCI America Supplier No. B32915G

Chemical Identifiers

CAS 151257-01-1
Molecular Formula C11H19ClN2O
Molecular Weight (g/mol) 230.736
MDL Number MFCD03428357
InChI Key WWRHZLCKSVQRBG-UHFFFAOYSA-N
PubChem CID 10176995
IUPAC Name 2-butyl-1,3-diazaspiro[4.4]non-1-en-4-one;hydrochloride
SMILES CCCCC1=NC2(CCCC2)C(=O)N1.Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 230.74
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Butyl-1,3-diazaspiro[4.4]non-1-en-4-one Hydrochloride