accessibility menu, dialog, popup

UserName

CAS RN 15121-89-8

CH3OOO(E/Z)

CAS RN 15121-89-8

IUPAC Name: ethyl 4-oxo-4-phenylbut-2-enoate
Synonyms: ethyl 4-oxo-4-phenylbut-2-enoate
Molecular Weight (g/mol): 204.23
Molecular Formula: C12H12O3
InChi Key: ACXLBHHUHSJENU-UHFFFAOYSA-N
SMILES: CCOC(=O)C=CC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results