accessibility menu, dialog, popup

UserName

CAS RN 151-73-5

IUPAC Name: disodium (1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-2,9a,11a-trimethyl-1-[2-(phosphonatooxy)acetyl]-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: betamethasone sodium phosphate betamethasone sodium phosphate [usan:ban:jan] betamethazone disodium phosphate
Molecular Weight (g/mol): 516.41
Molecular Formula: C22H28FNa2O8P
InChi Key: PLCQGRYPOISRTQ-LWCNAHDDSA-L
SMILES: [Na+].[Na+].C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP([O-])([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (3)
Grade 
  • (3)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)