missing translation for 'onlineSavingsMsg'
Learn More

Betamethasone 21-Phosphate Disodium Salt 98.0+%, TCI America™

Catalog No. B41101G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B41101G Supplier TCI America Supplier No. B41101G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 151-73-5
Molecular Formula C22H30FNa2O8P
Molecular Weight (g/mol) 518.426
MDL Number MFCD00200361
InChI Key ODEOTKLZVXBMJD-LWCNAHDDSA-N
PubChem CID 67151996
IUPAC Name [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] dihydrogen phosphate;sodium
SMILES CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)COP(=O)(O)O)O)C)O)F)C.[Na].[Na]

Specifications

Color White
Quantity 1 g
Formula Weight 516.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Betamethasone 21-Phosphate Disodium Salt
EINECSNumber (9)-1520
RTECSNumber TU4056500
TSCA No