accessibility menu, dialog, popup

UserName

H3C(R)CH3(S)NH2OHO

CAS RN 1509-34-8

IUPAC Name: (2S,3R)-2-amino-3-methylpentanoic acid
Synonyms: allo-isoleucine
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: AGPKZVBTJJNPAG-BJNJOQLTNA-N
SMILES: CC[C@@H](C)[C@H](N)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)