missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenyl Phosphorodichloridate 97.0+%, TCI America™

Catalog No. C09765G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C09765G Supplier TCI America Supplier No. C09765G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 15074-54-1
Color Yellow
Boiling Point 256°C
Molecular Formula C6H4Cl3O2P
Quantity 5 g
MDL Number MFCD00009704
UN Number 3265
Synonym 2-chlorophenyl phosphorodichloridate, 2-chlorophenyl dichlorophosphate, o-chlorophenyl dichlorophosphate, phosphorodichloridic acid, 2-chlorophenyl ester, o-chlorophenyl phosphorodichloridate, 2-chlorophenyl phosphoric acid dichloride, 2-chlorophenyl chlorophosphonochloridate, acmc-20anxn, o-chlorophenylphosphonyl dichloride, 2-chlorophenyl dichloridophosphate #
InChI Key VLDPXPPHXDGHEW-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)OP(=O)(Cl)Cl)Cl
IUPAC Name 1-chloro-2-dichlorophosphoryloxybenzene
Molecular Weight (g/mol) 245.42
PubChem CID 84775
Formula Weight 245.42
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Chlorophenyl Phosphorodichloridate [Phosphorylating Agent]
Show More Show Less