missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(chloroformyl)butyrate, 98%

Catalog No. AC126540100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC126540100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC126540100 Supplier Thermo Scientific Chemicals Supplier No. 126540100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1501-26-4 | C6H9ClO3 | 164.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

CAS 1501-26-4
Density 1.1910g/mL
Boiling Point 110.0°C (17.0 mmHg)
Flash Point 82°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C6H9ClO3
Refractive Index 1.4435 to 1.4455
Linear Formula ClCO(CH2)3CO2CH3
Quantity 10 g
MDL Number MFCD00000756
Specific Gravity 1.191
Synonym methyl 4-chloroformyl butyrate, methyl 5-chloro-5-oxovalerate, methyl 4-chloroformylbutyrate, methyl5-chloro-5-oxopentanoate, 5-chloro-5-oxopentanoic acid methyl ester, methyl glutaryl chloride, pentanoic acid, 5-chloro-5-oxo-, methyl ester, 4-chlorocarbonyl-butyric acid methyl ester, zlchem 559, pubchem14033
InChI Key JCAZSWWHFJVFPP-UHFFFAOYSA-N
SMILES COC(=O)CCCC(Cl)=O
IUPAC Name methyl 5-chloro-5-oxopentanoate
Molecular Weight (g/mol) 164.59
PubChem CID 73916
Formula Weight 164.59
Percent Purity 98%
Chemical Name or Material Methyl 4-(chloroformyl)butyrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately
EINECSNumber 216-115-6

RUO – Research Use Only