accessibility menu, dialog, popup

UserName

CAS RN 150-84-5

IUPAC Name: 3,7-dimethyloct-6-en-1-yl acetate
Synonyms: citronellyl acetate (S)-isomer citronellyl acetate citronellyl ethanoate (s)-citronellyl acetate (s)-isomer citronellyl acetate natural rhodinol, acetylated acetylated natural rhodinol acetic acid, citronellyl ester citronellol acetate ctronellyl acetate (natural)
Molecular Weight (g/mol): 198.31
Molecular Formula: C12H22O2
InChi Key: JOZKFWLRHCDGJA-UHFFFAOYNA-N
SMILES: CC(CCOC(C)=O)CCC=C(C)C
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)