missing translation for 'onlineSavingsMsg'
Learn More

Citronellyl acetate, 96%

Catalog No. AAA1987222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1987222 100 g
AAA1987214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1987222 Supplier Thermo Scientific Chemicals Supplier No. A1987222

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 150-84-5 | C12H22O2 | 198.306 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 150-84-5
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00015039
InChI Key JOZKFWLRHCDGJA-UHFFFAOYSA-N
Synonym citronellyl acetate, citronellol acetate, 3,7-dimethyl-6-octen-1-yl acetate, 3,7-dimethyloct-6-en-1-yl acetate, 1-acetoxy-3,7-dimethyloct-6-ene, acetic acid, citronellyl ester, natural rhodinol, acetylated, citronellyl ethanoate, b-citronellyl acetate, 6-octen-1-ol, 3,7-dimethyl-, acetate
PubChem CID 9017
ChEBI CHEBI:70478
IUPAC Name 3,7-dimethyloct-6-enyl acetate
SMILES CC(CCC=C(C)C)CCOC(=O)C

Specifications

Density 0.89
Boiling Point 240°C
Flash Point 98°C (208°F)
Refractive Index 1.4434
Quantity 100 g
Formula Weight 198.3
Percent Purity 96%
Chemical Name or Material Citronellyl acetate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 205-775-0
RTECSNumber RH3422500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only