missing translation for 'onlineSavingsMsg'
Learn More

Citronellyl Acetate 95.0+%, TCI America™

Catalog No. C091525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C091525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C091525ML Supplier TCI America Supplier No. C091525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 150-84-5
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00015039
InChI Key JOZKFWLRHCDGJA-UHFFFAOYSA-N
Synonym citronellyl acetate, citronellol acetate, 3,7-dimethyl-6-octen-1-yl acetate, 3,7-dimethyloct-6-en-1-yl acetate, 1-acetoxy-3,7-dimethyloct-6-ene, acetic acid, citronellyl ester, natural rhodinol, acetylated, citronellyl ethanoate, b-citronellyl acetate, 6-octen-1-ol, 3,7-dimethyl-, acetate
PubChem CID 9017
ChEBI CHEBI:70478
IUPAC Name 3,7-dimethyloct-6-enyl acetate
SMILES CC(CCC=C(C)C)CCOC(=O)C

Specifications

Color Colorless
Boiling Point 173°C
Quantity 25 mL
Formula Weight 198.31
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Citronellyl Acetate
RTECSNumber RH3422500
TSCA Yes