accessibility menu, dialog, popup

UserName

CAS RN 150-77-6

IUPAC Name: [2-(diethylazaniumyl)ethyl]diethylazanium
Synonyms: [2-(diethylammonio)ethyl]diethylazanium
Molecular Weight (g/mol): 174.33
Molecular Formula: C10H26N2
InChi Key: DIHKMUNUGQVFES-UHFFFAOYSA-P
SMILES: CC[NH+](CC)CC[NH+](CC)CC
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)