missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetraethylethylenediamine, 99+%

Catalog No. AC155590100
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC155590100 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC155590100 Supplier Thermo Scientific Chemicals Supplier No. 155590100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 150-77-6 | C10H26N2 | 174.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 150-77-6
Molecular Formula C10H26N2
Molecular Weight (g/mol) 174.33
MDL Number MFCD00009055
InChI Key DIHKMUNUGQVFES-UHFFFAOYSA-P
Synonym n,n,n',n'-tetraethylethylenediamine, teeda, 1,2-ethanediamine, n,n,n',n'-tetraethyl, tetraethylethylenediamine, ethylenediamine, n,n,n',n'-tetraethyl, 1,2-ethanediamine,n1,n1,n2,n2-tetraethyl, n,n,n',n'-tetraethylenediamine, 1,2-ethanediamine, n1,n1,n2,n2-tetraethyl, 2-diethylamino ethyl diethylamine, acmc-20ak9v
PubChem CID 67423
SMILES CC[NH+](CC)CC[NH+](CC)CC

Specifications

Color Yellow
Density 0.8080g/mL
Boiling Point 189.0°C to 192.0°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4333 to 1.4353
Linear Formula (C2H5)2NCH2CH2N(C2H5)2
Quantity 10 mL
Beilstein 04,251
Specific Gravity 0.808
Solubility Information Solubility in water: soluble.
IUPAC Name N,N,N',N'-tetraethylethane-1,2-diamine
Formula Weight 172.31
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material N,N,N',N'-Tetraethylethylenediamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 205-770-3

RUO – Research Use Only